Search
Search

CDC7| Gene Symbol: CDC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542756.2 | XP_005542813.2 | 2 - 3 | 153 | 105 | |
| XM_005542757.2 | XP_005542814.1 | 2 - 3 | 247 | 105 | ||
| XM_005542758.2 | XP_005542815.1 | 2 - 3 | 280 | 105 | ||
| XM_015432922.1 | XP_015288408.1 | 2 - 3 | 228 | 105 | ||
| XM_015432929.1 | XP_015288415.1 | 2 - 3 | 153 | 105 | ||
Set Membership: |
|