Search
Search

PSD3| Gene Symbol: PSD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562705.2 | XP_005562762.1 | 12 - 13 | 2581 | 72 | |
| XM_005562707.2 | XP_005562764.1 | 11 - 12 | 2666 | 72 | ||
| XM_005562708.2 | XP_005562765.1 | 9 - 10 | 1576 | 72 | ||
| XM_015454522.1 | XP_015310008.1 | 16 - 17 | 3700 | 72 | ||
| XM_015454523.1 | XP_015310009.1 | 13 - 14 | 2742 | 72 | ||
| XM_015454524.1 | XP_015310010.1 | 11 - 12 | 4069 | 72 | ||
| GenBank mRNA | AB055285.1 | - | 4 - 5 | 604 | 72 | |
| AB056822.1 | - | 4 - 5 | 641 | 72 | ||
| AB060909.1 | - | 4 - 5 | 639 | 72 | ||
Set Membership: |
|