Search
Search

DENND4A| Gene Symbol: DENND4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559835.2 | XP_005559892.1 | 18 - 19 | 2946 | 99 | |
| XM_015452965.1 | XP_015308451.1 | 13 - 14 | 2971 | 99 | ||
| XM_015452967.1 | XP_015308453.1 | 13 - 14 | 2971 | 99 | ||
| XM_015452968.1 | XP_015308454.1 | 13 - 14 | 2970 | 99 | ||
| XM_015452969.1 | XP_015308455.1 | 13 - 14 | 2970 | 99 | ||
Set Membership: |
|