Search
Search

SLC22A17| Gene Symbol: SLC22A17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560859.2 | XP_005560916.1 | 6 - 7 | 3405 | 94 | |
| XM_005560860.2 | XP_005560917.1 | 6 - 7 | 1378 | 94 | ||
| XM_015453585.1 | XP_015309071.1 | 6 - 7 | 3402 | 94 | ||
| XM_015453586.1 | XP_015309072.1 | 5 - 6 | 950 | 94 | ||
Set Membership: |
|