Search
Search

ATP13A1| Gene Symbol: ATP13A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588559.2 | XP_005588616.1 | 19 - 20 | 3055 | 122 | |
| XM_005588560.2 | XP_005588617.1 | 19 - 20 | 2674 | 122 | ||
| XM_005588561.2 | XP_005588618.1 | 19 - 20 | 3009 | 122 | ||
Set Membership: |
|