Search
Search

SNX4| Gene Symbol: SNX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547921.2 | XP_005547978.1 | 7 - 8 | 755 | 86 | |
| XM_005547922.2 | XP_005547979.1 | 7 - 8 | 800 | 86 | ||
| XM_015446322.1 | XP_015301808.1 | 7 - 8 | 649 | 86 | ||
| XM_015446323.1 | XP_015301809.1 | 6 - 7 | 632 | 86 | ||
| GenBank mRNA | AB172721.1 | - | 6 - 7 | 496 | 86 | |
| AB173524.1 | - | 6 - 7 | 628 | 86 | ||
Set Membership: |
|