Search
Search

VWA5B1| Gene Symbol: VWA5B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544555.2 | XP_005544612.1 | 2 - 3 | 331 | 101 | |
| XM_015441497.1 | XP_015296983.1 | 2 - 3 | 341 | 101 | ||
Set Membership: |
|