Search
Search

CDKL1| Gene Symbol: CDKL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015453765.1 | XP_015309251.1 | 2 - 3 | 685 | 134 | |
| XM_015453766.1 | XP_015309252.1 | 2 - 3 | 685 | 134 | ||
| XM_015453767.1 | XP_015309253.1 | 1 - 2 | 979 | 134 | ||
Set Membership: |
|