Search
Search

NUP214| Gene Symbol: NUP214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005580615.2 | XP_005580672.1 | 24 - 25 | 3568 | 62 | |
| XM_005580616.2 | XP_005580673.1 | 24 - 25 | 3564 | 62 | ||
| XM_015436318.1 | XP_015291804.1 | 24 - 25 | 3244 | 62 | ||
| XM_015436319.1 | XP_015291805.1 | 24 - 25 | 3568 | 62 | ||
Set Membership: |
|