Search
Search

RAD54B| Gene Symbol: RAD54B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563711.1 | XP_005563768.1 | 9 - 10 | 1648 | 103 | |
| XM_005563712.2 | XP_005563769.1 | 7 - 8 | 1671 | 103 | ||
| GenBank mRNA | AB168179.1 | - | 5 - 6 | 792 | 103 | |
Set Membership: |
|