Search
Search

ZNF777| Gene Symbol: ZNF777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551101.2 | XP_005551158.1 | 2 - 3 | 1030 | 79 | |
| XM_005551102.2 | XP_005551159.1 | 2 - 3 | 1566 | 79 | ||
| XM_015448243.1 | XP_015303729.1 | 1 - 2 | 4303 | 79 | ||
Set Membership: |
|