Search
Search

BOD1L1| Gene Symbol: BOD1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554493.2 | XP_005554550.1 | 10 - 11 | 8086 | 135 | |
| XM_005554494.2 | XP_005554551.1 | 10 - 11 | 8086 | 135 | ||
| XM_015450173.1 | XP_015305659.1 | 10 - 11 | 8086 | 135 | ||
| XM_015450174.1 | XP_015305660.1 | 10 - 11 | 8086 | 135 | ||
| XM_015450175.1 | XP_015305661.1 | 10 - 11 | 8086 | 135 | ||
| XM_015450176.1 | XP_015305662.1 | 10 - 11 | 10106 | 135 | ||
| XM_015450177.1 | XP_015305663.1 | 10 - 11 | 8086 | 135 | ||
Set Membership: |
|