Search
Search

RAD9B| Gene Symbol: RAD9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015431520.1 | XP_015287006.1 | 5 - 6 | 579 | 146 | |
| XM_015431521.1 | XP_015287007.1 | 5 - 6 | 579 | 146 | ||
| XM_015431522.1 | XP_015287008.1 | 5 - 6 | 579 | 146 | ||
| XM_015431523.1 | XP_015287009.1 | 4 - 5 | 497 | 146 | ||
| XM_015431524.1 | XP_015287010.1 | 5 - 6 | 563 | 146 | ||
| XM_015431525.1 | XP_015287011.1 | 5 - 6 | 559 | 146 | ||
| XM_015431526.1 | XP_015287012.1 | 4 - 5 | 472 | 146 | ||
| XM_015431527.1 | XP_015287013.1 | 4 - 5 | 463 | 146 | ||
Set Membership: |
|