Search
Search

CYP4F8| Gene Symbol: CYP4F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588293.2 | XP_005588350.1 | 4 - 5 | 466 | 116 | |
| XM_015440680.1 | XP_015296166.1 | 3 - 4 | 406 | 116 | ||
Set Membership: |
|