Search
Search

COPS2| Gene Symbol: COPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559512.2 | XP_005559569.1 | 13 | 1985 | 115 | |
| XM_005559513.2 | XP_005559570.1 | 13 | 1960 | 115 | ||
| GenBank mRNA | AB169606.1 | - | 13 | 1939 | 115 | |
Set Membership: |
|