Search
Search

KIFAP3| Gene Symbol: KIFAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005539954.2 | XP_005540011.1 | 18 - 19 | 2315 | 67 | |
| XM_005539956.2 | XP_005540013.1 | 18 - 19 | 2486 | 67 | ||
| XM_005539957.2 | XP_005540014.1 | 18 - 19 | 2315 | 67 | ||
| XM_005539960.2 | XP_005540017.1 | 16 - 17 | 4471 | 67 | ||
Set Membership: |
|