Search
Search

PEX14| Gene Symbol: PEX14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285002.1 | NP_001271931.1 | 8 - 9 | 687 | 95 | |
| XM_005544821.2 | XP_005544878.1 | 8 - 9 | 660 | 95 | ||
| XM_005544823.2 | XP_005544880.1 | 8 - 9 | 848 | 95 | ||
| GenBank mRNA | AB171677.1 | - | 8 - 9 | 687 | 95 | |
Set Membership: |
|