Search
Search

CR1| Gene Symbol: CR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015449302.1 | XP_015304788.1 | 28 - 29 | 6216 | 91 | |
| XM_015449307.1 | XP_015304793.1 | 28 - 29 | 6216 | 91 | ||
| XM_015449309.1 | XP_015304795.1 | 27 - 28 | 6074 | 91 | ||
| XM_015449313.1 | XP_015304799.1 | 28 - 29 | 5483 | 91 | ||
| XM_015449318.1 | XP_015304804.1 | 21 - 22 | 5271 | 91 | ||
| XM_015449320.1 | XP_015304806.1 | 28 - 29 | 5484 | 91 | ||