Search
Search

RIMBP2| Gene Symbol: RIMBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572618.2 | XP_005572675.1 | 8 - 9 | 2174 | 73 | |
| XM_005572622.2 | XP_005572679.1 | 13 - 14 | 3254 | 73 | ||
| XM_015431818.1 | XP_015287304.1 | 14 - 15 | 2711 | 73 | ||
| XM_015431819.1 | XP_015287305.1 | 13 - 14 | 4838 | 73 | ||
| XM_015431820.1 | XP_015287306.1 | 12 - 13 | 2487 | 73 | ||
| XM_015431821.1 | XP_015287307.1 | 13 - 14 | 2578 | 73 | ||
| XM_015431822.1 | XP_015287308.1 | 13 - 14 | 2577 | 73 | ||
| XM_015431823.1 | XP_015287309.1 | 13 - 14 | 2578 | 73 | ||
| XM_015431824.1 | XP_015287310.1 | 13 - 14 | 2572 | 73 | ||
| XM_015431825.1 | XP_015287311.1 | 13 - 14 | 2573 | 73 | ||