Search
Search

LOC102123898| Gene Symbol: LOC102123898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015442007.1 | XP_015297493.1 | 1 - 2 | 178 | 99 | |
| XM_015442008.1 | XP_015297494.1 | 1 - 2 | 178 | 99 | ||
| XM_015442013.1 | XP_015297499.1 | 1 - 2 | 177 | 99 | ||
| XM_015442014.1 | XP_015297500.1 | 1 - 2 | 185 | 99 | ||
Set Membership: |
|