Search
Search
| Gene Symbol: Rgl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009059.2 | NP_033085.2 | 8 - 9 | 1321 | 84 | |
| GenBank mRNA | AK144440.1 | - | 8 - 9 | 1323 | 84 | |
| AK215408.1 | - | 1 - 2 | 107 | 84 | ||
| BC068121.1 | - | 8 - 9 | 1321 | 6415928 | 84 | |
| KJ650210.1 | - | 7 - 8 | 1129 | 84 | ||
| U54639.1 | - | 8 - 9 | 1250 | 84 | ||
Set Membership: |
|