Search
Search
| Gene Symbol: Snx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019727.2 | NP_062701.2 | 7 - 8 | 843 | 103 | |
| GenBank mRNA | AB019214.1 | - | 7 - 8 | 783 | 103 | |
| AF154120.1 | - | 7 - 8 | 847 | 103 | ||
| AK149850.1 | - | 7 - 8 | 755 | 103 | ||
| AK154079.1 | - | 7 - 8 | 756 | 103 | ||
| AK155746.1 | - | 7 - 8 | 731 | 103 | ||
| AK161241.1 | - | 7 - 8 | 824 | 103 | ||
| AK167177.1 | - | 7 - 8 | 798 | 103 | ||
| AK167993.1 | - | 7 - 8 | 738 | 103 | ||
| AK168742.1 | - | 7 - 8 | 836 | 103 | ||
| BC066044.1 | - | 7 - 8 | 800 | 6807363 | 103 | |
| BC066189.1 | - | 7 - 8 | 734 | 30471747 | 103 | |
Set Membership: |
|