Search
Search
| Gene Symbol: Tlk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112705.2 | NP_001106176.1 | 16 - 17 | 1981 | 116 | |
| NM_001294329.1 | NP_001281258.1 | 14 - 15 | 1841 | 116 | ||
| NM_001294330.1 | NP_001281259.1 | 14 - 15 | 1789 | 116 | ||
| NM_001294331.1 | NP_001281260.1 | 16 - 17 | 1994 | 116 | ||
| NM_001294334.1 | NP_001281263.1 | 16 - 17 | 1981 | 116 | ||
| GenBank mRNA | AF045252.1 | - | 16 - 17 | 2000 | 116 | |
| AF045253.1 | - | 15 - 16 | 1428 | 116 | ||
| AF045254.1 | - | 11 - 12 | 1142 | 116 | ||
| AK014829.1 | - | 16 - 17 | 1619 | 116 | ||
| AK089925.1 | - | 14 - 15 | 1369 | 116 | ||
| AK165667.1 | - | 16 - 17 | 1984 | 116 | ||
| AX775743.1 | - | 10 - 11 | 1006 | 116 | ||
| BC066198.1 | - | 14 - 15 | 1334 | 116 | ||
| U56909.1 | - | 7 - 8 | 679 | 116 | ||
Set Membership: |
|