Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145620.4 | NP_663595.1 | 5 - 6 | 431 | 74 | |
GenBank mRNA | AF368232.1 | - | 5 - 6 | 414 | 74 | |
AK076113.1 | - | 5 - 6 | 433 | 74 | ||
AK156199.1 | - | 5 - 6 | 423 | 74 | ||
BC014703.1 | - | 5 - 6 | 431 | 4237895 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |