Search
Search

Capn3| Gene Symbol: Capn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109761.2 | NP_001103231.1 | 15 - 16 | 1790 | 67 | |
| NM_001177799.1 | NP_001171270.1 | 15 - 16 | 2057 | 67 | ||
| NM_007601.3 | NP_031627.2 | 18 - 19 | 2333 | 67 | ||
| GenBank mRNA | AF091998.1 | - | 15 - 16 | 1754 | 67 | |
| AF127766.2 | - | 15 - 16 | 1777 | 67 | ||
| AK049994.1 | - | 5 - 6 | 219 | 67 | ||
| AK088613.1 | - | 3 - 4 | 544 | 67 | ||
| BC090661.1 | - | 15 - 16 | 1790 | 30681055 | 67 | |
| BC139790.1 | - | 18 - 19 | 2053 | 40047690 | 67 | |
| X92523.1 | - | 18 - 19 | 2053 | 67 | ||
Set Membership: |
|