Search
Search

Gata3| Gene Symbol: Gata3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008091.3 | NP_032117.1 | 3 - 4 | 1368 | 52 | |
| GenBank mRNA | AK156634.1 | - | 3 - 4 | 1355 | 52 | |
| AK157625.1 | - | 988 | 52 | |||
| BC062915.1 | - | 2 - 3 | 964 | 6826352 | 52 | |
| X55123.1 | - | 2 - 3 | 984 | 52 | ||
Set Membership: |
|