Search
Search

Acot12| Gene Symbol: Acot12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028790.3 | NP_083066.1 | 4 - 5 | 370 | 93 | |
| GenBank mRNA | AB078618.1 | - | 4 - 5 | 367 | 93 | |
| AK004905.1 | - | 4 - 5 | 371 | 93 | ||
| AK033663.1 | - | 4 - 5 | 370 | 93 | ||
| AK034622.1 | - | 4 - 5 | 374 | 93 | ||
| BC025852.1 | - | 4 - 5 | 323 | 93 | ||
| BC132050.1 | - | 4 - 5 | 401 | 40130327 | 93 | |
| BC137996.1 | - | 4 - 5 | 401 | 8861015 | 93 | |
Set Membership: |
|