Search
Search
| Gene Symbol: Cd81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133655.2 | NP_598416.1 | 5 - 6 | 710 | 64 | |
| GenBank mRNA | AK136370.1 | - | 5 - 6 | 674 | 64 | |
| AK166521.1 | - | 5 - 6 | 676 | 64 | ||
| AK170741.1 | - | 5 - 6 | 675 | 64 | ||
| BC011433.1 | - | 5 - 6 | 665 | 2811801 | 64 | |
| X59047.1 | - | 5 - 6 | 430 | 64 | ||
Set Membership: |
|