Search
Search
| Gene Symbol: Add3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164099.2 | NP_001157571.1 | 7 - 8 | 1151 | 101 | |
| NM_001164100.2 | NP_001157572.1 | 7 - 8 | 1213 | 101 | ||
| NM_001164101.2 | NP_001157573.1 | 7 - 8 | 1151 | 101 | ||
| NM_001277100.1 | NP_001264029.1 | 7 - 8 | 1213 | 101 | ||
| NM_013758.4 | NP_038786.2 | 8 - 9 | 963 | 101 | ||
| GenBank mRNA | AF100424.1 | - | 6 - 7 | 882 | 101 | |
| AF100425.1 | - | 6 - 7 | 882 | 101 | ||
| AK019491.1 | - | 7 - 8 | 1092 | 101 | ||
| AK028481.1 | - | 7 - 8 | 1056 | 101 | ||
| AK035151.1 | - | 7 - 8 | 980 | 101 | ||
| AK083939.1 | - | 7 - 8 | 1054 | 101 | ||
| AK144174.1 | - | 7 - 8 | 1157 | 101 | ||
| BC004032.1 | - | 7 - 8 | 930 | 101 | ||
| BC029196.1 | - | 7 - 8 | 1016 | 101 | ||
| BC037116.1 | - | 8 - 9 | 974 | 4506748 | 101 | |
| BC060097.1 | - | 7 - 8 | 995 | 101 | ||
Set Membership: |
|