| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400018269.mm.1 |
NM_001164082.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018269.mm.1 |
NM_001164082.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018269.mm.1 |
NM_025945.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018269.mm.1 |
NM_025945.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018269.mm.1 |
TC1400002341.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400018271.mm.1 |
NM_001164082.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018271.mm.1 |
NM_001164082.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018271.mm.1 |
NM_025945.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018271.mm.1 |
NM_025945.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018271.mm.1 |
TC1400002341.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400018270.mm.1 |
NM_001164082.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018270.mm.1 |
NM_001164082.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018270.mm.1 |
NM_025945.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018270.mm.1 |
NM_025945.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018270.mm.1 |
TC1400002341.mm.1 |
MTA TC |