Search
Search

Prss44| Gene Symbol: Prss44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148940.3 | NP_683742.2 | 1 - 2 | 352 | 79 | |
| GenBank mRNA | AB047758.1 | - | 319 | 79 | ||
| AB162857.1 | - | 1 - 2 | 338 | 79 | ||
| AK006612.1 | - | 1 - 2 | 352 | 79 | ||
Set Membership: |
|