Search
Search
| Gene Symbol: Fbn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007993.2 | NP_032019.2 | 18 - 19 | 2459 | 97 | |
| GenBank mRNA | AF007248.1 | - | 2173 | 97 | ||
| L29454.1 | - | 4546 | 97 | |||
| U19972.1 | - | 17 - 18 | 2099 | 97 | ||
| U22493.1 | - | 18 - 19 | 2392 | 97 | ||
Set Membership: |
|