Search
Search
| Gene Symbol: Nit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023175.1 | NP_075664.1 | 6 - 7 | 554 | 62 | |
| GenBank mRNA | AF284573.1 | - | 6 - 7 | 554 | 62 | |
| AK003604.1 | - | 3 - 4 | 249 | 62 | ||
| AK004533.1 | - | 6 - 7 | 538 | 62 | ||
| AK004535.1 | - | 6 - 7 | 540 | 62 | ||
| BC020153.1 | - | 6 - 7 | 543 | 3992930 | 62 | |
Set Membership: |
|