Search
Search

Acsbg1| Gene Symbol: Acsbg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053178.2 | NP_444408.1 | 12 - 13 | 2048 | 75 | |
| GenBank mRNA | AB050554.1 | - | 12 - 13 | 2048 | 75 | |
| AK054103.1 | - | 12 - 13 | 1861 | 75 | ||
| AK129179.1 | - | 12 - 13 | 1844 | 75 | ||
| BC057322.1 | - | 12 - 13 | 1844 | 6832261 | 75 | |
Set Membership: |
|