Search
Search
| Gene Symbol: Clk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007713.4 | NP_031739.3 | 1 - 2 | 787 | 53 | |
| GenBank mRNA | AK090215.1 | - | 1 - 2 | 28 | 53 | |
| AK133936.1 | - | 1 - 2 | 33 | 53 | ||
| AK136481.1 | - | 1 - 2 | 50 | 53 | ||
| AK140015.1 | - | 1 - 2 | 30 | 53 | ||
| AK145880.1 | - | 1 - 2 | 42 | 53 | ||
| AK146942.1 | - | 1 - 2 | 790 | 53 | ||
| AK154003.1 | - | 1 - 2 | 36 | 53 | ||
| AK167300.1 | - | 1 - 2 | 789 | 53 | ||
| BC085084.1 | - | 1 - 2 | 30 | 6309020 | 53 | |
Set Membership: |
|