Search
Search

Ccser2| Gene Symbol: Ccser2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027045.1 | NP_081321.1 | 6 - 7 | 2324 | 91 | |
| NM_028407.3 | NP_082683.2 | 4 - 5 | 496 | 91 | ||
| GenBank mRNA | AK005931.1 | - | 4 - 5 | 496 | 91 | |
| AK129295.1 | - | 5 - 6 | 1896 | 91 | ||
| AK147383.1 | - | 6 - 7 | 2319 | 91 | ||
| AK172153.1 | - | 6 - 7 | 2298 | 91 | ||
| BC048641.1 | - | 4 - 5 | 495 | 91 | ||
| BC060947.1 | - | 4 - 5 | 495 | 91 | ||
| BC111465.1 | - | 5 - 6 | 705 | 91 | ||
| BC139090.1 | - | 5 - 6 | 2112 | 8862112 | 91 | |
Set Membership: |
|