Search
Search

Ppp2r5e| Gene Symbol: Ppp2r5e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012024.2 | NP_036154.1 | 4 - 5 | 1025 | 86 | |
| GenBank mRNA | AK053228.1 | - | 3 - 4 | 875 | 86 | |
| AK088366.1 | - | 4 - 5 | 997 | 86 | ||
| AK132432.1 | - | 4 - 5 | 1058 | 86 | ||
| AK220163.1 | - | 4 - 5 | 1015 | 86 | ||
| BC085149.1 | - | 4 - 5 | 1025 | 6848033 | 86 | |
| BC094387.1 | - | 4 - 5 | 1017 | 86 | ||
| U49728.1 | - | 2 - 3 | 302 | 87 | ||
Set Membership: |
|