| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32246208 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32246209 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32246205 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32246204 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32246207 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32246203 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32246206 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2078815 |
AY534248.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2078815 |
AF155511.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2078815 |
NM_023500.2 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P6828 |
BP764926.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P6828 |
BC054851.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P6828 |
NM_023500.2 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P6828 |
BP764926.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P6828 |
BC054851.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P6828 |
NM_023500.2 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
969853 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
502466 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
240837 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
255472 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
330925 |
AK017897.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
330925 |
AK153408.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
330925 |
BC118516.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
330925 |
XM_001472888.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
330925 |
XM_001473721.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
330925 |
XM_203312.6 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
330925 |
XM_896683.3 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
330925 |
XM_896707.2 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
330925 |
XM_920202.3 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
330925 |
XM_920212.2 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
330925 |
XM_920225.2 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
632798 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
634755 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
135028 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
170143 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
885769 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
753289 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
56555 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
19988 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
175289 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P176349 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P118109 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2078815 |
AY534248.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2078815 |
AF155511.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2078815 |
NM_023500.2 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P6828 |
BP764926.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P6828 |
BC054851.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P6828 |
NM_023500.2 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P6828 |
BP764926.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P6828 |
BC054851.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P6828 |
NM_023500.2 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P123371 |
BC125620.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P123371 |
BC125618.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P123371 |
AY534248.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P123371 |
BC027097.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P123371 |
AF155511.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P315861 |
AY534248.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P315861 |
AF155511.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246201 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246208 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246198 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246170 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246172 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246180 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246202 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246181 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246179 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246195 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246196 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246211 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246189 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246186 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246194 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246206 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246200 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246173 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246191 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246210 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246205 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246199 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246185 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246204 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246177 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246174 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246184 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246187 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246197 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32246193 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03636690 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09229773 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09229742 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03636688 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03636691 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P176349 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P118109 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P123371 |
BC125620.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P123371 |
BC125618.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P123371 |
AY534248.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P123371 |
BC027097.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P123371 |
AF155511.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P315861 |
AY534248.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P315861 |
AF155511.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P6828 |
BP764926.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P6828 |
BC054851.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P6828 |
NM_023500.2 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P6828 |
BP764926.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P6828 |
BC054851.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P6828 |
NM_023500.2 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P58240 |
AY534248.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P58240 |
AF155511.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P58240 |
NM_023500.2 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P133087 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P176349 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P09229773 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P118109 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14608969 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14608964 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14608965 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14920223 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32246203 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32246206 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32246204 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32246207 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32246205 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32246209 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32246208 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246398 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246185 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246189 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246403 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246374 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246378 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246380 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246409 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246193 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246440 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246431 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246359 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246375 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246423 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246244 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246430 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246396 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246207 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246383 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246411 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246202 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246201 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246474 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246384 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246421 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246181 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246415 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246387 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246376 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246191 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246179 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246184 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246187 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246211 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246173 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246413 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246424 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246373 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246170 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246406 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246425 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246381 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246447 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246386 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246407 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246172 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246197 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246343 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246435 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246365 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246444 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246177 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246369 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246439 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246442 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246427 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246416 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246200 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246366 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246256 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246307 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246418 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246180 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246344 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246438 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246390 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246443 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246437 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246414 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246434 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246198 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246370 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246205 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246399 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246196 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246393 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246199 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246254 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246371 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32246208 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14608965 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246187 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246387 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14723185 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246198 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246179 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246196 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246170 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246197 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246210 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246189 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246202 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246394 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246344 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246384 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246174 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246303 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246185 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246388 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246193 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14723161 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246390 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246184 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246186 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14920223 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246379 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246346 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246312 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246173 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246211 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246343 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246177 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246393 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246172 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246391 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246381 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246194 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246168 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246386 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246307 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246200 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32246181 |
- |
- |