Search
Search
| Gene Symbol: Cfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142706.1 | NP_001136178.1 | 1 - 2 | 470 | 64 | |
| NM_008198.2 | NP_032224.2 | 1 - 2 | 470 | 64 | ||
| GenBank mRNA | AK098094.1 | - | 1 - 2 | 130 | 64 | |
| AK149531.1 | - | 1 - 2 | 103 | 64 | ||
| AK149573.1 | - | 1 - 2 | 129 | 64 | ||
| AK151246.1 | - | 1 - 2 | 204 | 64 | ||
| BC005451.1 | - | 1 - 2 | 104 | 3591669 | 64 | |
| CA492475.1 | - | 1 - 2 | 507 | 64 | ||
| M57890.1 | - | 1 - 2 | 173 | 64 | ||
Set Membership: |
|