| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000011646.mm.1 |
NM_001136082.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011646.mm.1 |
NM_001136082.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011646.mm.1 |
NM_001164080.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011646.mm.1 |
NM_001164080.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011646.mm.1 |
NM_001164081.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011646.mm.1 |
NM_001164081.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011646.mm.1 |
NM_011589.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011646.mm.1 |
NM_011589.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011646.mm.1 |
TC1000001593.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000011647.mm.1 |
NM_001136082.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011647.mm.1 |
NM_001136082.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011647.mm.1 |
NM_001164080.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011647.mm.1 |
NM_001164080.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011647.mm.1 |
NM_001164081.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011647.mm.1 |
NM_001164081.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011647.mm.1 |
NM_011589.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011647.mm.1 |
NM_011589.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011647.mm.1 |
TC1000001593.mm.1 |
MTA TC |