| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800002519.mm.1 |
NM_175375.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002519.mm.1 |
NM_175375.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002519.mm.1 |
TC1800000332.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800002521.mm.1 |
NM_175375.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002521.mm.1 |
NM_175375.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002521.mm.1 |
TC1800000332.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800002522.mm.1 |
NM_175375.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002522.mm.1 |
NM_175375.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002522.mm.1 |
TC1800000332.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800002520.mm.1 |
NM_175375.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002520.mm.1 |
NM_175375.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002520.mm.1 |
TC1800000332.mm.1 |
MTA TC |