Search
Search

Grik4| Gene Symbol: Grik4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175481.5 | NP_780690.2 | 18 - 19 | 2468 | 67 | |
| GenBank mRNA | AK032029.1 | - | 18 - 19 | 2469 | 67 | |
| BC118010.2 | - | 17 - 18 | 2383 | 40096690 | 67 | |
Set Membership: |
|