Search
Search

Tspan15| Gene Symbol: Tspan15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197996.2 | NP_932113.2 | 5 - 6 | 669 | 67 | |
| GenBank mRNA | AK012471.1 | - | 5 - 6 | 669 | 66 | |
| CN455615.1 | - | 5 - 6 | 558 | 67 | ||
| CX206586.1 | - | 5 - 6 | 671 | 67 | ||
Set Membership: |
|