Search
Search

Ppil6| Gene Symbol: Ppil6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028430.1 | NP_082706.1 | 2 - 3 | 283 | 75 | |
| GenBank mRNA | AK013818.1 | - | 2 - 3 | 246 | 75 | |
| BC147756.1 | - | 2 - 3 | 279 | 9056705 | 75 | |
| BC147757.1 | - | 2 - 3 | 279 | 9056706 | 75 | |
| CA466046.1 | - | 2 - 3 | 265 | 75 | ||
Set Membership: |
|