Search
Search

Plxna3| Gene Symbol: Plxna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008883.2 | NP_032909.2 | 1 - 2 | 152 | 62 | |
| GenBank mRNA | BC093482.1 | - | 1 - 2 | 135 | 30945372 | 62 |
| CJ126001.1 | - | 1 - 2 | 153 | 62 | ||
Set Membership: |
|