| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000002710.mm.1 |
NM_001168289.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002710.mm.1 |
NM_001168289.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002710.mm.1 |
NM_027772.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002710.mm.1 |
NM_027772.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002710.mm.1 |
TC1000000392.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000002707.mm.1 |
NM_001168289.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002707.mm.1 |
NM_001168289.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002707.mm.1 |
NM_027772.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002707.mm.1 |
NM_027772.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002707.mm.1 |
TC1000000392.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000002711.mm.1 |
NM_001168289.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002711.mm.1 |
NM_001168289.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002711.mm.1 |
NM_027772.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002711.mm.1 |
NM_027772.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002711.mm.1 |
TC1000000392.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000002713.mm.1 |
NM_001168289.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002713.mm.1 |
NM_001168289.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002713.mm.1 |
NM_027772.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002713.mm.1 |
NM_027772.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002713.mm.1 |
TC1000000392.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000002709.mm.1 |
NM_001168289.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002709.mm.1 |
NM_001168289.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002709.mm.1 |
NM_027772.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002709.mm.1 |
NM_027772.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002709.mm.1 |
TC1000000392.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000002712.mm.1 |
NM_001168289.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002712.mm.1 |
NM_001168289.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002712.mm.1 |
NM_027772.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002712.mm.1 |
NM_027772.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002712.mm.1 |
TC1000000392.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000002708.mm.1 |
NM_001168289.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002708.mm.1 |
NM_001168289.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002708.mm.1 |
NM_027772.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002708.mm.1 |
NM_027772.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000002708.mm.1 |
TC1000000392.mm.1 |
MTA TC |