Search
Search
| Gene Symbol: Tcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130458.1 | NP_001123930.1 | 7 - 8 | 1143 | 114 | |
| NM_001130459.1 | NP_001123931.1 | 7 - 8 | 1112 | 114 | ||
| NM_015749.3 | NP_056564.1 | 7 - 8 | 1146 | 114 | ||
| GenBank mRNA | AF090686.1 | - | 6 - 7 | 1073 | 114 | |
| AK133707.1 | - | 7 - 8 | 1108 | 114 | ||
| AK143761.1 | - | 7 - 8 | 1114 | 114 | ||
| AK147176.1 | - | 7 - 8 | 1117 | 114 | ||
| AK161618.1 | - | 7 - 8 | 1113 | 114 | ||
| AK170401.1 | - | 7 - 8 | 1132 | 114 | ||
| AK186678.1 | - | 1 - 2 | 64 | 114 | ||
| AK216881.1 | - | 1 - 2 | 161 | 114 | ||
| BC003720.1 | - | 6 - 7 | 1019 | 3601531 | 114 | |
Set Membership: |
|