Search
Search

Klkb1| Gene Symbol: Klkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008455.2 | NP_032481.2 | 9 - 10 | 1124 | 94 | |
| GenBank mRNA | BC026555.1 | - | 9 - 10 | 1050 | 4972788 | 94 |
Set Membership: |
|