Search
Search

Tnrc18| Gene Symbol: Tnrc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122730.2 | NP_001116202.2 | 9 - 10 | 3368 | 62 | |
| NM_178242.2 | NP_839973.1 | 9 - 10 | 3368 | 62 | ||
| GenBank mRNA | AK141849.1 | - | 6 - 7 | 2465 | 62 | |
| AK173280.1 | - | 6 - 7 | 1126 | 62 | ||
| BC025631.1 | - | 4 - 5 | 901 | 62 | ||
| BC049818.1 | - | 9 - 10 | 3368 | 6511384 | 62 | |
Set Membership: |
|