Search
Search
| Gene Symbol: Tfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289507.1 | NP_001276436.1 | 8 - 9 | 1296 | 94 | |
| NM_001289509.1 | NP_001276438.1 | 10 - 11 | 1315 | 94 | ||
| NM_001289511.1 | NP_001276440.1 | 9 - 10 | 1173 | 94 | ||
| NM_015799.4 | NP_056614.3 | 10 - 11 | 1255 | 94 | ||
| GenBank mRNA | AF207741.1 | - | 8 - 9 | 1130 | 94 | |
| AF222895.1 | - | 9 - 10 | 1173 | 94 | ||
| AK004848.2 | - | 10 - 11 | 1231 | 94 | ||
| AK004965.1 | - | 10 - 11 | 1286 | 94 | ||
| AK155526.1 | - | 9 - 10 | 1393 | 94 | ||
| AK165320.1 | - | 8 - 9 | 1124 | 94 | ||
| BC013654.1 | - | 8 - 9 | 1296 | 4196597 | 94 | |
Set Membership: |
|